Learn More
Medchemexpress LLC Navitoclax-piperazine | 2143096-93-7 | 99.7% | 973.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY444325MG
Encompass_Preferred
Navitoclax-piperazine is an analog of Navitoclax and a BCL-XL inhibitor. It acts as a ligand for the target protein of PROTAC DT2216. Navitoclax-piperazine and E3 ubiquitin ligase VHL ligand can be used to synthesize PROTAC DT2216.
  • Is an analog of Navitoclax and a BCL-XL inhibitor.
  • Is the ligand for the target protein of PROTAC DT2216.
  • Can be used with E3 ubiquitin ligase VHL ligand to synthesize PROTAC DT2216.
  • For research use only.
  • Appearance: solid, white to off-white.
Catalog No. 50-004-03529