Learn More
Medchemexpress LLC Navitoclax-piperazine | 2143096-93-7 | MFCD32693949 | 99.7% | 973.63 | C47H56ClF3N6O5S3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4443210MG
Encompass_Preferred
Navitoclax-piperazine is a research chemical and analog of navitoclax that functions as a BCL-XL inhibitor, supplied as a white to off-white solid for biochemical and cell-based studies. It is typically provided in small milligram quantities and is characterized by high purity and a large molecular weight.
  • BCL-XL inhibitor useful for apoptosis and oncology research.
  • High purity (~99.7%) suitable for research applications.
  • Supplied as a white to off-white solid for ease of handling.
  • Available in small milligram quantities and as 10 mM solutions in DMSO for ready use.
  • Molecular formula C47H56ClF3N6O5S3; molecular weight 973.63.
Catalog No. 50-002-98789