Learn More
Medchemexpress LLC Navlimetostat hydrochloride | 00-00-0 | MFCD34604075 | 99.2% | 501.34 g/mol | C23H19Cl2FN6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY139611C5MG
Encompass_Preferred
Navlimetostat hydrochloride is a small-molecule research inhibitor that selectively targets the PRMT5-MTA complex. It shows potent biochemical inhibition and antineoplastic activity in vitro and in vivo, and is supplied as a solid for laboratory research use.
  • Selective PRMT5-MTA complex inhibitor for research applications.
  • Potent activity with low-nanomolar inhibitory values in biochemical assays.
  • Supplied as a solid with defined molecular formula and weight.
  • High purity appropriate for biochemical and cellular studies.
  • Storage recommendations provided for both solid and solvent forms.
  • Available in small research sizes for preclinical experiments.
Catalog No. 50-002-23060