Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naz2329 | 2809469-05-2 | 99.0% | 501.56 g/mol | C21H18F3NO4S3 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10369325MG

Encompass_Preferred

NAZ2329 is a small-molecule inhibitor of PTPRZ and PTPRG intended for research use. It is supplied as a 25 mg research sample and is characterized by high purity and defined physicochemical properties suitable for biochemical and in vivo studies.

  • Inhibits PTPRZ and PTPRG activity.
  • High purity: 99.02%.
  • Molecular formula: C21H18F3NO4S3; molecular weight: 501.56 g/mol.
  • Soluble in DMSO; in vitro solubility reported at 100 mg/mL.
  • Provided as a 25 mg sample suitable for preliminary biochemical and in vivo assays.

Catalog No. 50-002-45705


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.