Learn More
Medchemexpress LLC HY-12872 5mg Medchemexpress, Nazartinib CAS:1508250-71-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128725MG
Encompass_Preferred
Medchemexpress, HY-12872 5mg Nazartinib CAS:1508250-71-2 Nazartinib (EGF816) is a novel, covalent mutant-selective EGFR inhibitor, with Ki and Kinact of 31 nM and 0.222 min−1 on EGFR(L858R/790M) mutant, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0124