Learn More
Medchemexpress LLC NB-598 | 131060-14-5 | 449.67 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16343
Encompass_Preferred
NB-598 is a potent and competitive inhibitor of squalene epoxidase (SE) that suppresses triglyceride biosynthesis through the farnesol pathway. This compound also functions as a click chemistry reagent, featuring an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Potent and competitive inhibitor of squalene epoxidase
  • Suppresses triglyceride biosynthesis
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Suitable for immunology and inflammation research pathways
  • Powder storage at -20°C for 3 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-94747