Learn More
Medchemexpress LLC Nbd-14270 | 2411819-82-2 | 99.5% | 425.43 | C18H18F3N5O2S | 5 MG

Supplier: Medchemexpress LLC HY1399895MG
NBD-14270 is a pyridine analogue that functions as a potent HIV-1 entry antagonist by binding to the gp120 envelope protein. It exhibits sub-micromolar antiviral activity with low cytotoxicity and is provided for research applications with characterized purity, formula, and storage recommendations.
- High purity: 99.53%.
- Chemical formula: C18H18F3N5O2S; molecular weight: 425.43.
- CAS number: 2411819-82-2.
- Recommended solvent: DMSO; use ultrasonic aid for dissolution.
- Storage: solid at 4°C; in solvent store at -80°C for long-term stability.
- Typical applications: in vitro antiviral and HIV-1 entry assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.