Learn More
Biotium NBD C6-sphingomyelin
Supplier:  Biotium 60031
Encompass_Preferred
NBD C6-sphingomyelin (6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosyl phosphocholine) has been used for following sphingolipid metabolism in cells.Properties: Ex/Em (MeOH) = 466/536 nm; Dark solid soluble in chloroform or DMSO; Store at -20°C and protect from light; C35H61N6O9P; MW: 740.87; [94885-04-8].
Catalog No. 50-196-4726