Learn More
Cayman Chemical NBD-FTY720 phenoxy hydrochlo
SDP
Supplier:  Cayman Chemical 16841500 UG
Encompass_Preferred
A fluorescently-labeled analog of FTY720; methanol side chain of FTY720 that is targeted for phosphorylation by sphingosine kinases is retained, suggesting that it would likewise be phosphorylated in vivo
Catalog No. 50-296-5292