Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NBD-Pen | 1955505-54-0 | 99.9% | 390.46 | C19H28N5O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1291095MG

Encompass_Preferred

NBD-Pen is a turn-on fluorescent probe for selective detection of lipid-derived radicals in biological systems. It enables live-cell imaging and biochemical studies of lipid peroxidation and oxidative stress with sensitive fluorescence readout.

  • High selectivity and sensitivity for lipid radicals.
  • Turn-on fluorescence enables low-background detection in cells.
  • Excitation at 470 nm and emission at 530 nm for standard imaging.
  • High purity (99.94%) suitable for research use.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Store protected from light; follow solvent storage guidelines.

Catalog No. 50-002-63119


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.