Learn More
Medchemexpress LLC NBD Sphingosine | 1449370-25-5 | ≥99.0% | 50 UG
SDP
Supplier:  Medchemexpress LLC HY13043350UG
Encompass_Preferred
NBD Sphingosine (NBD-Sph) is a fluorochrome, a fluorescence-labeled sphingosine. It can be used for fluorescence assays for sphingosine kinases.
  • Used for fluorescence assays for sphingosine kinases
  • Fluorochrome
  • Fluorescence-labeled sphingosine
Catalog No. 50-003-40245