Learn More
Cayman Chemical NBenzylacetamIdInhydrobro 10mg
SDP
Supplier:  Cayman Chemical 1357010 MG
Encompass_Preferred
A potent inhibitor of iNOS (IC50 = 0.20 μM), with over 1,000-fold selectivity compared to eNOS (IC50 = 350 μM)
Catalog No. 50-295-5831