Learn More
Enzo Life Sciences NBQX (5mg). CAS: 118876-58-7
SDP
Supplier:  Enzo Life Sciences ALX550044M005
Encompass_Preferred
Most potent and selective AMPA/kainate receptor antagonist. For the more soluble form see NBQX . disodium salt (ALX-550-272). Alternative name: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline, 6-Nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione. Purity: ≥99% (HPLC). Appearance: Yellow crystalline powder. Solubility: Soluble in DMSO. A 1mM aqueous solution may be prepared by taking up in >2.5 eq. of NaOH and backtitrating to pH 8.0-8.5 with conc. HCl. Long Term Storage: +4°C.
Catalog No. 50-200-9466