Learn More
Abcam NBQ x disodium salt, AMPA / kainate antagonist, 50MG
Supplier:  Abcam AB12004650MG
Encompass_Preferred
MW 380.25 Da, Purity >98%. Water soluble, potent, selective and competitive AMPA/kainate receptor antagonist. Neuroprotective and anticonvulsant in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4716