Learn More
Abcam NBQ x disodium salt (mM/ml), AMPA / kainate antagonist, 50MM_ML
Supplier:  Abcam AB14448950MM_ML
Encompass_Preferred
MW 380.25 Da, Purity >98%. Water soluble, potent, selective and competitive AMPA/kainate receptor antagonist. Neuroprotective and anticonvulsant in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5671