Learn More
Cayman Chemical NButyldeoxynojIrImycInhydr 5mg
SDP
Supplier:  Cayman Chemical 210655 MG
Encompass_Preferred
An inhibitor of UDP-glucose ceramide glucosyltransferase and β-glucosidase 2 (IC50s = 32 and 81 μM, respectively, for the rat recombinant enzymes); increases the activity of wild-type acid β-glucosidase, as well as the S364R, N370S, V15M, or M123T mutants, in COS-7 cells expressing the human enzymes at 10 µM; inhibits HIV-1 and HIV-2 infection of PBMCs (IC50s = 282 and 211 μM, respectively)
Catalog No. 50-298-3887