Learn More
Cayman Chemical NbutyrylLHomoserine lactone
SDP
Supplier:  Cayman Chemical 1000789810 MG
Encompass_Preferred
A bacterial quorum sensing signal molecule; regulates virulence in general, infection prevention, and formation of biofilms
Catalog No. 50-288-8610