Learn More
TARGETMOL CHEMICALS INC NCGC00092410 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4054410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCGC00092410 (ML008) is a glucocerebrosidase (GC) inhibitor with IC50 value of 31 nM. It can be used for the study of Gaucher disease caused by mutation of glucocerebrase gene. purity: 98%
Catalog No. 50-257-9983