Learn More
ATCC NCI-H2085 (CRL-5921) Human Lung Adenocarcinoma (NSCLC) Cell Line
Supplier: ATCC CRL5921
NCI-H2085 (CRL-5921) is a human non-small cell lung adenocarcinoma cell line established from a 45-year-old male donor. It is used in lung cancer research, including cell biology, genomics, and preclinical drug testing.
- Species: Homo sapiens.
- Tissue: Lung.
- Disease: Adenocarcinoma (non-small cell lung cancer).
- Donor sex and age: Male, 45 years.
- Growth medium: ACL-4 base; complete medium requires 5% fetal bovine serum.
- Year of origin: 1988.
- Depositors: AF Gazdar, JD Minna.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.