Learn More
ATCC NCI-H929-GFP-Luc2 (CRL-3580-GFP-LUC2) Human plasmacytoma cell line, GFP and Luc2 reporters, suspension
Supplier:  ATCC CRL3580GFPLUC2
Encompass
NCI-H929-GFP-Luc2 is a human plasmacytoma (myeloma) suspension cell line engineered to constitutively express green fluorescent protein (GFP) and firefly luciferase (luc2). It is intended for use as a target cell in immuno-oncology research, CAR-T cytotoxicity assays, and cell-based assay development.
  • Human plasmacytoma (bone marrow) cell line.
  • Expresses green fluorescent protein and firefly luciferase (luc2).
  • Suitable as a target for CAR-T and immuno-oncology assays.
  • Suspension growth suitable for cell-based killing assays.
  • Recommended medium: RPMI-1640 with 10% fetal bovine serum and 0.05 mM 2-mercaptoethanol.
  • Maintained under puromycin selection (0.06 μg/mL) to retain reporter expression.
  • Cryopreserved frozen vials; store in vapor phase of liquid nitrogen.
Catalog No. NC3901755