Learn More
ATCC NCI-H1650 (H1650); Bronchoalveolar carcinoma; Human (Homo Sapiens)
Supplier:  ATCC CRL5883
Encompass
NCI-H1650 [H-1650, H1650] is a cell line exhibiting epithelial morphology that was isolated in 1987 from the lung tissue of a 27-year-old, male smoker with stage 3B, bronchoalveolar carcinoma. Use these cells for your immuno-oncology and lung cancer research.
Catalog No. 50-238-4139