Learn More
ATCC NCI-H1703; Lung Squamous; cell carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5889
Encompass
NCI-H1703 [H1703] is a squamous cell line exhibiting epithelial morphology that was isolated from the lungs of a 54-year-old, White male patient with non-small cell lung cancer. This product has applications in cancer research.
Catalog No. 50-238-1783
 Restricted Item