Learn More
ATCC NCI-H1869; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5900
Encompass
NCI-H1869 [H1869] are sqamous cells isolated from the lungs of a 58-year-old, White male patient with stage 4 non-small cell lung cancer. This product has applications in cancer research.
Catalog No. 50-238-3164
 Restricted Item