Learn More
ATCC NCI-H2066; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5917
Encompass
NCI-H2066 [H2066] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 70-year-old, White, female patient with stage 1 small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-3088