Learn More
ATCC NCI-H2087; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5922
Encompass
NCI-H2087 [H2087] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 69-year-old, White, male patient with stage 1 non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-1886