Learn More
ATCC NCI-H226; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5826
Encompass
NCI-H226 [H226] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a male with squamous cell carcinoma. This product has applications for cancer research and toxicology.
Catalog No. 50-238-3899