Learn More
ATCC NCI-H2347; Lung Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL5942
Encompass
NCI-H2347 [H2347] is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 54-year-old, White female patient with stage 1 non-small cell lung cancer: adenocarcinoma. This product has applications in cancer research.
Catalog No. 50-238-3863