Learn More
ATCC NCI-H441; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRMHTB174
Encompass
NCI-H441 [H441] is an epithelial-like cell that was isolated from the lung of a male with papillary adenocarcinoma. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-2202