Learn More
ATCC NCI-H716 (H716); Colon Cancer; Human (Homo Sapies)
Supplier:  ATCC CCL251
Encompass
NCI-H716 [H716] is a cell line exhibiting lymphoblast morphology that was isolated in 1984 from the cecum of a White, 33-year-old, male patient with colorectal adenocarcinoma. This cell line was deposited by AF Gazdar and can be used in cancer research.
Catalog No. 50-238-4142