Learn More
ATCC NCI-H820; Lung Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB181
Encompass
NCI-H820 [H820] is an epithelial-like cell that was isolated from the lung of a 53-year-old, White, male with papillary adenocarcinoma.
Catalog No. 50-238-2458