Learn More
Cayman Chemical ANTIBODY CRnAD 2 25mg
SDP
Supplier:  Cayman Chemical 1981425 MG
Encompass_Preferred
An affinity probe for Aβ; has high affinity for Aβ (Kd = 38 nM); exhibits a 70-fold increase in fluorescence intensity, a blue shift from 805 to 715 nm, and a large increase in quantum yield when bound to Aβ aggregates
Catalog No. 50-298-2103