Learn More
Medchemexpress LLC Nd-336 | 1807453-83-3 | MFCD34469331 | 97.2% | 371.90 g·mol⁻¹ | C16H18ClNO3S2 | 500UG

Supplier: Medchemexpress LLC HY124373500UG
ND-336 is a small-molecule matrix metalloproteinase (MMP) inhibitor (CAS 1807453-83-3) used for preclinical research. It selectively inhibits MMP-2, MMP-9, and MMP-14 and has been reported to accelerate diabetic wound healing in animal models. The compound has molecular formula C16H18ClNO3S2, molecular weight ~371.90 g·mol⁻¹, and listed purity around 97.2%.
- Selective inhibitor of MMP-2, MMP-9, and MMP-14.
- Reported to accelerate diabetic wound healing in vivo.
- Molecular formula C16H18ClNO3S2 and molecular weight ~371.90 g·mol⁻¹.
- High listed purity of approximately 97.2% suitable for research use.
- Long-term storage recommended at -20°C; in solution store at -80°C for extended stability.
- Intended for in vitro and in vivo research applications; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.