Learn More
Cayman Chemical NDesmethyltamoxIfenhydroch 5mg
SDP
Supplier:  Cayman Chemical 406735 MG
Encompass_Preferred
An active metabolite of tamoxifen; inhibits PKC (IC50 = 8 µM for the rat enzyme); decreases iron (III) chloride and ascorbate-induced increases in TBARS in rat liver microsomes (IC50 = 17.5 µM); induces relaxation in isolated and washed rat aortic rings (pD2 = 9)
Catalog No. 50-294-3080