Learn More
Med Vet International BD PrecisionGlide Needles, 19G x 1.5 in, 100/box
Supplier:  Med Vet International 305187
Encompass_Preferred
This BD 305187 Needle is a 19G x 1.5" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-304-3645