Learn More
Med Vet International AHS Needle, 30 X 1/2in, Hypodermic, 1000/BX, AHS3013RN
Supplier:  Med Vet International AHS3013RNX10
Encompass_Preferred
This AHS AHS3013RN Needle is a 30 X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/BX
Catalog No. 50-118-0463