Learn More
Cayman Chemical ndrosteron 250mg
SDP
Supplier:  Cayman Chemical 15872250 MG
Encompass_Preferred
A steroid hormone produced in the body from 5α-reduced metabolites of dehydroepiandrosterone that possesses 7-fold weaker androgen potency compared to that of testosterone; excreted in urine and used as the international reference standard for androgenic activity
Catalog No. 50-297-1794