Learn More
Gold Biotechnology Inc NDSB-201 (Non-detergent sulfobetaine) 250 g
SDP
Supplier:  Gold Biotechnology Inc N201250
Encompass
NDSB-201 is a protein specific additive that can be used to help solubilize and stabilize proteins at non-denaturing concentrations while preventing protein aggregation.
Catalog No. NC1528982