Learn More
FUJIFILM BIOSCIENCES INC NDSB-256
Supplier:  FUJIFILM BIOSCIENCES INC 14907962
Encompass_Preferred
This product is a non-surfactant with zwitterionic character It is used for the extraction of membrane proteins and nuclear proteins the refolding of denatured proteins and the crystallization of proteins NDSB-256 is zwitterionic compound which have a quaternary ammonium ion acidic and hydrophilic sulfonate and short hydrophobic group in one molecule Such compound that has hydrophilic and hydrophobic groups in the molecule typically form micelle and is called as surfactant but NDSB-256 does not form micelle due to those short hydrophobic groups Readily removed by dialysis as micelle is not formed Keep zwitterionic character in a wide range of pH For research use only RUO
Catalog No. 50-004-50285