Learn More
ABclonal Technology NDUFS3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22162100UL
Encompass_Preferred
This gene encodes one of the iron-sulfur protein (IP) components of mitochondrial NADH ubiquinone oxidoreductase (complex I) Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency
Catalog No. 50-000-07603