Learn More
Apexbio Technology LLC NE 100 hydrochloride 10mg
SDP
Supplier:  Apexbio Technology LLC B728110
Encompass_Preferred
NE 100 hydrochloride (CAS 149409-57-4) is a selective antagonist of the sigma-1 receptor exhibiting an IC50 value of 4 16 nM By targeting the sigma-1 receptor this small molecule has been shown to inhibit ER stress-induced hippocampal cell death and demonstrates antipsychotic activity in preclinical models NE 100 hydrochloride is widely utilized in basic neuroscience research to investigate the physiological and pathological roles of sigma-1 receptor signaling particularly in the context of neuropsychiatric disorders and neuroprotection
Catalog No. 50-000-88818