Learn More
Medchemexpress LLC Ne 52-qq57 | 1401728-56-0 | 99.9% | 416.52 g/mol | C24H28N6O | 10 MG

Supplier: Medchemexpress LLC HY10178410MG
NE 52-QQ57 is a selective, orally available antagonist of GPR4 with an IC50 of ~70 nM and reported anti-inflammatory activity; supplied as a high-purity research compound for in vitro and in vivo pharmacology studies.
- Selective GPR4 antagonist (IC50 70 nM).
- High purity (99.9%).
- Molecular weight 416.52 g/mol.
- Soluble in DMSO (20 mg/mL).
- Stable storage: powder at -20°C (up to 3 years).
- Demonstrated oral efficacy in animal models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.