Learn More
Medchemexpress LLC Nebentan potassium 5mg | 342005-82-7 | 5MG
SDP
Supplier:  Medchemexpress LLC HY106994A5MG
Encompass_Preferred
Nebentan potassium (YM598) is a potent selective and orally active non-peptide endothelin ETA receptor antagonist through the modification of Bosentan (HY-A0013) Nebentan potassium inhibits [125I endothelin-1 binding to cloned human endothelin ETA and ETB receptor with Ki of 0 697 nM and 569 nM respectively[1 YM598 can ameliorate the progression of cor pulmonale and myocardial infarction in vivo[2
Catalog No. 50-002-39878