Learn More
Selleck Chemical LLC Nebivolol hydrochloride (R-65824) S1549-50mg
SDP
Supplier:  Selleck Chemical LLC S154950MG
Encompass_Preferred
Nebivolol HCl (R-65824) selectively inhibits 1-adrenoceptor with IC50 of 0 8 nM
Catalog No. 50-311-0107