Learn More
New England Biolabs, Inc. NEBNext Enzymatic 5hmC-seq Kit 96 rxns
SDP
Supplier:  New England Biolabs, Inc. E3350L
Encompass
The NEBNext Enzymatic 5hmC-seq Kit specifically detects 5hmC sites. The kit includes NEBNext Ultra II library prep reagents, and 5hmC is detected using a two-step enzymatic conversion workflow that minimizes damage to DNA and allows discrimination of 5hmC from both cytosine and 5mC after Illumina sequencing. E5hmC-seq data can also be subtracted from EM-seq data, allowing determination of the precise location of individual 5mC and 5hmC sites. The kit includes the E5hmC-seq adaptor and index primers are supplied separately.
Catalog No. 50-255-1938