Learn More
New England Biolabs, Inc. NEBNext End Repair Module – 100 reactions
SDP
Supplier:  New England Biolabs, Inc. E6050L
Encompass
This module is part of the original standard library prep workflow, which is longer than the NEBNext Ultra II DNA Library Prep Kit workflow, has more components, and requires more clean-up steps. It also requires higher input amounts: 1 - 5 µg of input DNA vs. 500 pg - 1 µg required for the Ultra II DNA workflow.
Catalog No. 50-431-3