Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit v2 (Human/Mouse/Rat) – 24 reactions
SDP
Supplier:  New England Biolabs, Inc. E7400L
Encompass
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human, mouse and rat total RNA preparations. This product is suitable for both intact and degraded RNA (e.g. FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications.
Catalog No. 50-202-4021