Learn More
New England Biolabs, Inc. NEBNext Library Quant DNA Standards - 6 standards - 500 reactions
SDP
Supplier:  New England Biolabs, Inc. E7642S
Encompass
NEBNext Library Quant DNA Standards are a set of six standards defining a curve from 100 pM to 0.001 pM intended for use with the NEBNext Library Quant Master Mix found in the NEBNext Library Quant Kit for Illumina (NEB E7630). The six standards offered are the same as the six that are included in the base kit. The NEBNext Library Quant Kit manual includes details for using either four or six standards for quantitation. 500 reactions
Catalog No. 50-223-7901