Learn More
Medchemexpress LLC Neboglamine hydrochl 100mg | 2759182-59-5 | 292.80 g/mol | C13H25ClN2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114753A100MG
Encompass_Preferred
Neboglamine hydrochloride is the hydrochloride salt of neboglamine, an orally active positive allosteric modulator of the NMDA receptor glycine site used in neuroscience and schizophrenia research. The compound is supplied as a white to off-white solid and is available in multiple formats for in vitro and in vivo studies.
  • Orally active NMDA receptor glycine-site positive modulator.
  • Intended for neuroscience and schizophrenia research applications.
  • Supplied as a white to off-white solid hydrochloride salt for ease of handling.
  • Reported high purity suitable for research use.
  • Available in small-mass packs and ready-to-use DMSO solution formats for assays.
Catalog No. 50-002-29706