Learn More
TARGETMOL CHEMICALS INC Neboglamine hydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T606121MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Neboglamine (CR-2249, XY-2401) hydrochloride, an orally active positive modulator of the NMDA receptor glycine site, can be used in schizophrenia research [1]. Purity 99.11%
Catalog No. 50-287-6634