Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit (Bacteria) with RNA Sample Purification Beads – 24 reactions
SDP
Supplier:  New England Biolabs, Inc. E7860L
Encompass
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic). For convenience, this kit includes RNA Clean beads.
Catalog No. 50-202-4030